1,3-Diphenylisobenzofuran 97%
Supplier: Thermo Scientific Chemicals
14758-0010
14758-0050
TS14758-0010EA
39
USD
TS14758-0010
TS14758-0050
1,3-Diphenylisobenzofuran 97%
1,3-Diphenylisobenzofuran
Formula:
C₂₀H₁₄O Melting Pt: 129…133 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005931 CAS Number: 5471-63-6 |
Specification Test Results
Appearance | Yellow to green Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 128 to 137 °C |
GC | ≥96.0 % |
pH | 3.6 to 4.0 (0.5 % in water) |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...