Ethyl toluene-4-sulphonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 4-toluenesulphonate, Ethyl p-toluenesulphonate, p-Toluenesulphonic acid ethyl ester, Ethyl 4-methylbenzenesulphonate, Toluene-4-sulphonic acid ethyl ester

14723-0010 14723-0500 14723-2500
TS14723-0010EA 218 USD
TS14723-0010 TS14723-0500 TS14723-2500
Ethyl toluene-4-sulphonate 98%
Ethyl toluene-4-sulphonate
Formula: C₉H₁₂O₃S
MW: 200.26 g/mol
Boiling Pt: 158…162 °C (10 mmHg)
Melting Pt: 32…34 °C
Density: 1.174 g/cm³
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00009100
CAS Number: 80-40-0
EINECS: 201-276-7
Merck Index: 1303889

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5120 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR