Pentafluorobenzoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,3,4,5,6-Pentafluorobenzoyl chloride, Pentafluorobenzoylchloride, PFBC
14712-0050
14712-0250
14712-1000
TS14712-0050EA
70
USD
TS14712-0050
TS14712-0250
TS14712-1000
Pentafluorobenzoyl chloride 98%
Pentafluorobenzoyl chloride
Formula:
C₇ClF₅O MW: 230.52 g/mol Boiling Pt: 158 Density: 1.6 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00000657 CAS Number: 2251-50-5 EINECS: 218-842-4 UN: 3265 ADR: 8 II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4520 to 1.4540 (20°C, 589 nm) |
GC | ≥97.5 % |
Titration with HCl | 98.5 to 101.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...