Sudan I, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Scarlet B, 1-(Phenyldiazenyl)naphthalen-2-ol, 1-Phenylazo-2-naphthol, Oil Orange, Solvent Yellow 14, Sudan Yellow

14706-0250 14706-1000
TS14706-0250EA 41 USD
TS14706-0250 TS14706-1000
Sudan I, pure
Sudan I
Formula: C₁₆H₁₂N₂O
MW: 248.28 g/mol
Boiling Pt: 444 °C (1013 hPa)
Melting Pt: 131…133 °C
MDL Number: MFCD00003911
CAS Number: 842-07-9
EINECS: 212-668-2

Order Now

Specification Test Results

Appearance Orange to red to brown Powder
Infrared spectrum Conforms
UV Lambda max at approximately 418 and 476 nm (in ethanol)
Specific extinction E (1%/1cm) ≥500 (at 476 nm, in ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR