Catechol violet, pure indicator grade

Supplier: Thermo Scientific Chemicals

14654-0050 14654-0250
TS14654-0050EA 75 USD
TS14654-0050 TS14654-0250
Catechol violet, pure indicator grade
Catechol violet
Formula: C₁₉H₁₄O₇S
Storage Temperature: Ambient
MDL Number: MFCD00005868
CAS Number: 115-41-3
EINECS: 204-088-3

Order Now

Specification Test Results

Appearance Green to dark green to brown Powder
Infrared spectrum Conforms
UV spectrum Complies
Solubility (0.1 % in water) Clear
UV Lambda max 441 to 449 nm (in methanol)
Suitability (as complexom. indicator) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR