Trimethyl phosphonoacetate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Methyl 2-(dimethoxyphosphoryl)acetate, Dimethyl methoxycarbonylmethanephosphonate
14023-0250
14023-1000
14023-5000
TS14023-0250EA
51
USD
TS14023-0250
TS14023-1000
TS14023-5000
Trimethyl phosphonoacetate 98%
Trimethyl phosphonoacetate
Formula:
C₅H₁₁O₅P MW: 182.11 g/mol Boiling Pt: 265…268 °C Density: 1.26 g/cm³ Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008452 CAS Number: 5927-18-4 EINECS: 227-663-0 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4360 to 1.4380 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...