Pivaloyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

14005-0010 14005-0050 14005-2500
TS14005-0010EA 117 USD
TS14005-0010 TS14005-0050 TS14005-2500
Pivaloyl chloride 99%
Pivaloyl chloride
Formula: C₅H₉ClO
Boiling Pt: 105 °C (1013 hPa)
Melting Pt: –56 °C
Density: 0.98 g/cm³
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00000709
CAS Number: 3282-30-2
EINECS: 221-921-6
UN: 2438
ADR: 6.1,I

Order Now

Specification Test Results

Appearance Clear colorless to light pink Liquid
Infrared spectrum Conforms
Refractive index 1.4110 to 1.4130 (20°C, 589 nm)
GC ≥98.5 %
Titration RCOCl 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR