2',4',6'-Trihydroxyacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phloroacetophenone, THAP, 2-Acetylphloroglucinol

13979-0050 13979-0250 13979-1000
TS13979-0050EA 44 USD
TS13979-0050 TS13979-0250 TS13979-1000
2',4',6'-Trihydroxyacetophenone 98%
2',4',6'-Trihydroxyacetophenone
Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 219…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00149091
CAS Number: 480-66-0
EINECS: 207-556-5

Order Now

Specification Test Results

Appearance Yellow to orange to tan Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 217 to 222 °C
Water ≤10 % (K.F.)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR