Triethyl orthopropionate 98+%

Supplier: Thermo Scientific Chemicals
Warning

13967-0025 13967-1000 13967-5000
TS13967-0025EA 541 USD
TS13967-0025 TS13967-1000 TS13967-5000
Triethyl orthopropionate 98+%
Triethyl orthopropionate
Formula: C₉H₂₀O₃
MW: 176.26 g/mol
Boiling Pt: 155…160 °C (760 mmHg)
Melting Pt: < 0 °C
Density: 0.886 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00009226
CAS Number: 115-80-0
EINECS: 204-108-0
UN: 3271
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4010 to 1.4030 (20°C, 589 nm)
GC ≥98.0 %
Specific optical rotation +18° to +28° (20°C, 589 nm) (c=2,ethanol/0.5N HCl(4:1))

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR