Triethyl orthoformate 98%

Supplier: Thermo Scientific Chemicals
Warning

13966-0010 13966-0025 13966-0250 13966-1000
TS13966-0010EA 110 USD
TS13966-0010 TS13966-0025 TS13966-0250 TS13966-1000
Triethyl orthoformate 98%
Triethyl orthoformate
Formula: C₇H₁₆O₃
MW: 148.2 g/mol
Boiling Pt: 146 °C (760 mmHg)
Melting Pt: –61 °C
Density: 0.89 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009230
CAS Number: 122-51-0
EINECS: 204-550-4
UN: 2524
ADR: 3,III
Merck Index: 1207012

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3900 to 1.3920 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR