Tributylphosphine 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tri-n-butylphosphine, TBP

13934-1000 13934-5000
TS13934-1000EA 71 USD
TS13934-1000 TS13934-5000
Tributylphosphine 95%
Tributylphosphine
Formula: C₁₂H₂₇P
MW: 202.32 g/mol
Boiling Pt: 240…242 °C (760 mmHg)
Melting Pt: –65 °C
Density: 0.81 g/cm³
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00009462
CAS Number: 998-40-3
EINECS: 213-651-2
UN: 3254
ADR: 4.2,I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4630 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR