Thiophene 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

13882-0025 13882-1000 13882-5000
TS13882-0025EA 204 USD
TS13882-0025 TS13882-1000 TS13882-5000
Thiophene 99+%, Extra Pure
Thiophene
Formula: C₄H₄S
MW: 84.14 g/mol
Boiling Pt: 84 °C (760 mmHg)
Melting Pt: –38 °C
Density: 1.065 g/cm³
Flash Pt: –9 °C
Storage Temperature: Ambient
MDL Number: MFCD00005413
CAS Number: 110-02-1
EINECS: 203-729-4
UN: 2414
ADR: 3,II
Merck Index: 1209490

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5270 to 1.5290 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥99.0 %
Color scale ≤50 APHA
Benzene ≤0.1 % (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR