Sodium tetraphenylborate 99.5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt

13852-0250 13852-1000
TS13852-0250EA 135 USD
TS13852-0250 TS13852-1000
Sodium tetraphenylborate 99.5%
Sodium tetraphenylborate
Formula: (C₆H₅)₄BNa
MW: 342.22 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00011494
CAS Number: 143-66-8
EINECS: 205-605-5
UN: 2811
ADR: 6.1,III
Merck Index: 1308765

Order Now

Specification Test Results

Appearance White to light beige Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C)
Titration with HClO4 99.4 to 100.6 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR