Tetrachloro-o-benzoquinone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4,5,6-Tetrachloro-1,2-benzoquinone, Tetrachloro-1,2-benzoquinone, 2-Chloranil, Tetrachloro-o-benzoquinone, o-Chloranil, 3,4,5,6-Tetrachlorocyclohexa-3,5-diene-1,2-dione

13799-0050 13799-0250
TS13799-0050EA 69 USD
TS13799-0050 TS13799-0250
Tetrachloro-o-benzoquinone 97%
Tetrachloro-o-benzoquinone
Formula: C₆Cl₄O₂
MW: 245.88 g/mol
Boiling Pt: 312 °C (1013 hPa)
Melting Pt: 125…129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001646
CAS Number: 2435-53-2
EINECS: 219-424-4

Order Now

Specification Test Results

Appearance Bordeaux Crystalline powder
Infrared spectrum Conforms
Melting point 125 to 129 °C
GC ≥96.0 %
Specific optical rotation +12° to +15° (20 °C, 589 nm) (Neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR