Isopropyltriphenylphosphonium iodide 98%

Supplier: Thermo Scientific Chemicals
Warning

13708-0250 13708-1000
TS13708-0250EA 94 USD
TS13708-0250 TS13708-1000
Isopropyltriphenylphosphonium iodide 98%
Isopropyltriphenylphosphonium iodide
Formula: C₂₁H₂₂IP
Melting Pt: 192…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00031595
CAS Number: 24470-78-8
EINECS: 246-281-5

Order Now

Specification Test Results

Appearance Light yellow to beige to light brown Powder or crystals
Infrared spectrum Conforms
Melting point 195 to 200 °C
Titration Argentometric ≥97.5 %
Specific optical rotation +67° to +69° (20 °C, 589 nm) (c=2.3, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR