Semicarbazide hydrochloride 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Semicarbazide hydrochloride, Carbamylhydrazine hydrochloride, Aminourea hydrochloride, Hydrazinecarboxamide hydrochloride, N-Aminourea hydrochloride, Hydrazinecarboxamidehydrochloride

13264-0025 13264-1000 13264-5000
TS13264-0025EA 302 USD
TS13264-0025 TS13264-1000 TS13264-5000
Semicarbazide hydrochloride 99+%
Semicarbazide hydrochloride
Formula: CH₆ClN₃O
MW: 111.53 g/mol
Melting Pt: 170…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00013009
CAS Number: 563-41-7
EINECS: 209-247-0
UN: 2811
ADR: 6.1 III
Merck Index: 12,08588

Order Now

Specification Test Results

Appearance White Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 170 to 178 °C
Sulfated ash ≤0.1 %
Solubility (10 % in water) Clear colorless
Titration with KIO3 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR