1,10-Phenanthroline hydrochloride monohydrate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,10-Phenanthroline Chloride Monohydrate, 1,10-Phenanthrolinium chloride monohydrate, 1,10-Phenanthroline monohydrochloride monohydrate

13014-0100 13014-0250 13014-1000
TS13014-0100EA 86 USD
TS13014-0100 TS13014-0250 TS13014-1000
1,10-Phenanthroline hydrochloride monohydrate 97%
1,10-Phenanthroline hydrochloride monohydrate
Formula: C₁₂H₉ClN₂·1H₂O
Melting Pt: 224…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00150061
CAS Number: 18851-33-7
EINECS: 223-325-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow to light purple Powder or crystals
Infrared spectrum Conforms
Water 7 to 10 % (K.F.)
Sulfated ash ≤0.2 %
Titration with HClO4 ≥96.0 %
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.5 M soln.) Passes test
Suitability (HPLC gradient) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR