Nitro blue tetrazolium chloride monohydrate (NBT) 90%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride

12859-0010 12859-0050
TS12859-0010EA 204 USD
TS12859-0010 TS12859-0050
Nitro blue tetrazolium chloride monohydrate (NBT) 90%
Nitro blue tetrazolium chloride monohydrate (NBT)
Formula: C₄₀H₃₀Cl₂N₁₀O₆·H₂O·CH₄O
MW: 867.71 g/mol
Melting Pt: >170 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00150013
CAS Number: 298-83-9

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (105°C)
Sulfated ash ≤0.1 %
HPLC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR