2-Nitroaniline 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Nitroaniline

12835-0050 12835-0051 12835-1000 12835-5000
TS12835-0050EA 22 USD
TS12835-0050 TS12835-0051 TS12835-1000 TS12835-5000
2-Nitroaniline 98%
2-Nitroaniline
Formula: C₆H₆N₂O₂
Boiling Pt: 284 °C (760 mmHg)
Melting Pt: 70…74 °C
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00007687
CAS Number: 88-74-4
EINECS: 201-855-4
UN: 1661
ADR: 6.1,II
Merck Index: 1206680

Order Now

Specification Test Results

Appearance Orange to brown Crystals or flakes
Infrared spectrum Conforms
Melting point 70 to 74 °C
GC ≥97.5 %
Calcium (Ca) ≤80 ppm (Typical)
Chromium (Cr) ≤0.5 ppm (Typical)
Iron (Fe) ≤65 ppm (Typical)
Potassium (K) ≤50 ppm (Typical)
Titanium (Ti) ≤150 ppm (Typical)
Silicon dioxide (SiO2) ≥99.80 % (Typical)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR