α-Naphthoflavone 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Phenyl-4H-benzo[h]chromen-4-one, 2-Phenylbenzo[h]chromen-4-one, 7,8-Benzoflavone

12816-0010 12816-0050
TS12816-0010EA 36 USD
TS12816-0010 TS12816-0050
α-Naphthoflavone 97%
α-Naphthoflavone
Formula: C₁₉H₁₂O₂
MW: 272.3 g/mol
Boiling Pt: 431 °C (1013 hPa)
Melting Pt: 155…158 °C
MDL Number: MFCD00004985
CAS Number: 604-59-1
EINECS: 210-071-1

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 155 to 158 °C
HPLC ≥99.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR