2-Mesitylenesulphonyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,4,6-Trimethylbenzenesulphonyl chloride, Mesitylene-2-sulphonyl chloride

12559-0250 12559-1000 12559-5000
TS12559-0250EA 67 USD
TS12559-0250 TS12559-1000 TS12559-5000
2-Mesitylenesulphonyl chloride 99%
2-Mesitylenesulphonyl chloride
Formula: C₉H₁₁ClO₂S
MW: 218.7 g/mol
Boiling Pt: 150 °C (20 mmHg)
Melting Pt: 53…57 °C
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD00007434
CAS Number: 773-64-8
EINECS: 212-257-8
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to grey Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 57 °C
Titration Argentometric ≥98.5 % (ex Cl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR