2-Hydroxy-1,4-naphthoquinone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-p-naphthoquinone, 2-Hydroxy-1,4-naphoquinone

12163-0100 12163-0250 12163-1000
TS12163-0100EA 29 USD
TS12163-0100 TS12163-0250 TS12163-1000
2-Hydroxy-1,4-naphthoquinone 99%
2-Hydroxy-1,4-naphthoquinone
Formula: C₁₀H₆O₃
MW: 174.16 g/mol
Melting Pt: 192…195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00001678
CAS Number: 83-72-7
EINECS: 201-496-3

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR