Ethyl cyanoformate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11830-0100
11830-0500
TS11830-0100EA
65
USD
TS11830-0100
TS11830-0500
Ethyl cyanoformate 99%
Ethyl cyanoformate
Formula:
C₄H₅NO₂ Density: 1 g/cm³ Flash Pt: 24 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001836 CAS Number: 623-49-4 EINECS: 210-797-9 UN: 2929 ADR: 6.1,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3810 to 1.3830 (20°C, 589 nm) |
GC | ≥98.5 % |
Titration with HClO4 | 99.0 to 101.0 % |
Chloride (Cl) | ≤5 ppm |
Phosphate (PO4) | ≤5 ppm |
Sulfate (SO4) | ≤50 ppm |
Ammonium (NH4) | ≤30 ppm |
Calcium (Ca) | ≤100 ppm |
Iron (Fe) | ≤10 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...