Ethyl acetoacetate 99%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Acetoacetic acid ethyl ester, EAA

11797-0010 11797-0025 11797-0250
TS11797-0010EA 58 USD
TS11797-0010 TS11797-0025 TS11797-0250
Ethyl acetoacetate 99%, pure
Ethyl acetoacetate
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 180.8 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.03 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00009199
CAS Number: 141-97-9
EINECS: 205-516-1
Merck Index: 1303793

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4180 to 1.4200 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥98.5 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Sodium (Na) ≤0.05 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.01 %
Nitrogen compounds (as N) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR