Sanger's reagent 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11703-0050
11703-0250
11703-1000
11703-5000
TS11703-0050EA
48
USD
TS11703-0050
TS11703-0250
TS11703-1000
TS11703-5000
Sanger's reagent 98%
Sanger's reagent
Formula:
C₆H₃FN₂O₄ Boiling Pt: 296 °C Melting Pt: 23…26 °C Density: 1.48 g/cm³ Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007056 CAS Number: 70-34-8 EINECS: 200-734-3 UN: 2811 ADR: 6.1,II Merck Index: 1304199 |
Specification Test Results
Appearance | Yellow to brown Liquid or low melting solid |
Infrared spectrum | Conforms |
Refractive index | 1.5675 to 1.5705 (20°C, 589 nm) |
GC | ≥97.5 % |
Residue after ignition | ≤0.05 % |
Suitability | (for precip. of Fe, etc.) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...