Dansyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-(Dimethylamino)naphthalene-1-sulphonyl chloride

11585-0010 11585-0050 11585-0250
TS11585-0010EA 70 USD
TS11585-0010 TS11585-0050 TS11585-0250
Dansyl chloride 98%
Dansyl chloride
Formula: C₁₂H₁₂ClNO₂S
MW: 269.75 g/mol
Melting Pt: 71…74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003985
CAS Number: 605-65-2
EINECS: 210-092-6
UN: 3261
ADR: 8,II
Merck Index: 1302840

Order Now

Specification Test Results

Appearance Yellow to light orange Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 70 to 74 °C
HPLC ≥97.5 %
UV at 420 nm A: ≤0.01
Color scale ≤15 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR