4-(Dimethylamino)cinnamaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Dimethylamino-Cinnamaldehyde

11579-0050 11579-0250
TS11579-0050EA 87 USD
TS11579-0050 TS11579-0250
4-(Dimethylamino)cinnamaldehyde 98%
4-(Dimethylamino)cinnamaldehyde
Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
Melting Pt: 137…142 °C
Storage Temperature: Freezer
MDL Number: MFCD00007002
CAS Number: 6203-18-5
EINECS: 228-267-0

Order Now

Specification Test Results

Appearance Yellow to orange Powder
Infrared spectrum Conforms
Melting point 137 to 142 °C
Titration with HClO4 ≥97.5 %
Stabilizer 0.1 % Hydroquinone

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR