1,2-Diiodoethane 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11514-0250
11514-1000
TS11514-0250EA
66
USD
TS11514-0250
TS11514-1000
1,2-Diiodoethane 99%
1,2-Diiodoethane
Formula:
C₂H₄I₂ Boiling Pt: 206 °C (1013 hPa) Melting Pt: 80…83 °C Density: 2.13 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00001093 CAS Number: 624-73-7 EINECS: 210-859-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...