3,4-Dihydro-2H-pyran 99%

Supplier: Thermo Scientific Chemicals
Danger

11478-1000 11478-5000
TS11478-1000EA 43 USD
TS11478-1000 TS11478-5000
3,4-Dihydro-2H-pyran 99%
3,4-Dihydro-2H-pyran
Formula: C₅H₈O
MW: 84.12 g/mol
Melting Pt: –70 °C
Density: 0.92 g/cm³
Flash Pt: –9 °C
Storage Temperature: Ambient
MDL Number: MFCD00006558
CAS Number: 110-87-2
EINECS: 203-810-4
UN: 2376
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4395 to 1.4435 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR