Diethyl carbonate 99%

Supplier: Thermo Scientific Chemicals
Warning

11422-0010 11422-0025 11422-0100 11422-1000 11422-5000
TS11422-0010EA 84 USD
TS11422-0010 TS11422-0025 TS11422-0100 TS11422-1000 TS11422-5000
Diethyl carbonate 99%
Diethyl carbonate
Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 126…128 °C (760 mmHg)
Melting Pt: –43 °C
Density: 0.975 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00009107
CAS Number: 105-58-8
EINECS: 203-311-1
UN: 2366
ADR: 3 III
Merck Index: 1303815

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3830 to 1.3860 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer Approx. 0.01 % Citric acid
Relative density (20/4°C) 1.026 to 1.036
Acid value ≤5 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR