1,4-Diazabicyclo[2.2.2]octane 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DABCO

11247-0025 11247-0250 11247-1000 11247-5000
TS11247-0025EA 654 USD
TS11247-0025 TS11247-0250 TS11247-1000 TS11247-5000
1,4-Diazabicyclo[2.2.2]octane 97%
1,4-Diazabicyclo[2.2.2]octane
Formula: C₆H₁₂N₂
MW: 112.17 g/mol
Boiling Pt: 174
Melting Pt: 155…160 °C
Density: 1.14 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00006689
CAS Number: 280-57-9
EINECS: 205-999-9
UN: 1325
ADR: 4.1 II
Merck Index: 12,09801

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Water ≤2.0 % (K.F.)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR