3,3'-Diaminobenzidine tetrahydrochloride hydrate, (max. 10% H₂O) 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DAB HCl hydrate, 3,3',4,4'-Biphenyltetramine tetrahydrochloride hydrate, DAB tetrahydrochloride hydrate

11209-0010 11209-0050 11209-0250
TS11209-0010EA 39 USD
TS11209-0010 TS11209-0050 TS11209-0250
3,3'-Diaminobenzidine tetrahydrochloride hydrate, (max. 10% H₂O) 97%
3,3'-Diaminobenzidine tetrahydrochloride hydrate
Formula: C₁₂H₁₄N₄·4HCl·H₂O
MW: 360.11 g/mol (anhy)
Melting Pt: 300 °C
Storage Temperature: Freezer
MDL Number: MFCD08273058
CAS Number: 868272-85-9

Order Now

Specification Test Results

Appearance Grey to brown-purple Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 % (on dry substance)
Loss on drying ≤10 % (120°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR