3,3'-Diaminobenzidine (DAB) 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: [1,1'-Biphenyl]-3,3',4,4'-tetraamine, DAB, 3,3',4,4'-Biphenyltetramin, Biphenyl-3,3',4,4'-tetrayltetraamine

11208-0050 11208-0250
TS11208-0050EA 53 USD
TS11208-0050 TS11208-0250
3,3'-Diaminobenzidine (DAB) 98+%
3,3'-Diaminobenzidine (DAB)
Formula: C₁₂H₁₄N₄
MW: 214.27 g/mol
Melting Pt: 176…180 °C
Flash Pt: >200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007725
CAS Number: 91-95-2
EINECS: 202-110-6

Order Now

Specification Test Results

Appearance Off-white to brown Powder or crystals
Infrared spectrum Conforms
Melting point 175 to 179 °C
HPLC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR