2,4,6-Trichloro-1,3,5-triazine 99%

Supplier: Thermo Scientific Chemicals
Danger

11089-0010 11089-0051 11089-2500
TS11089-0010EA 72 USD
TS11089-0010 TS11089-0051 TS11089-2500
2,4,6-Trichloro-1,3,5-triazine 99%
2,4,6-Trichloro-1,3,5-triazine
Formula: C₃Cl₃N₃
Boiling Pt: 190 °C (760 mmHg)
Melting Pt: 145…148 °C
Density: 1.92 g/cm³
Flash Pt: >200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006046
CAS Number: 108-77-0
EINECS: 203-614-9
UN: 2670
ADR: 8,II

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 145 to 148 °C
Titration ≥98.5 %
Anticaking agent May contain 0% to 0.5% Silica as an additive


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR