Chlorodiphenylphosphine 95% tech.

Supplier: Thermo Scientific Chemicals
Danger

10913-0250 10913-1000 10913-5000
TS10913-0250EA 44 USD
TS10913-0250 TS10913-1000 TS10913-5000
Chlorodiphenylphosphine 95% tech.
Chlorodiphenylphosphine
Formula: C₁₂H₁₀ClP
Boiling Pt: 320 °C (760 mmHg)
Melting Pt: 14…16 °C
Density: 1.229 g/cm³
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00000529
CAS Number: 1079-66-9
EINECS: 214-093-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow Liquid (May develop some turbidity or precipitate)
Infrared spectrum Conforms
Refractive index 1.6340 to 1.6370 (20°C, 589 nm)
GC ≥94 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Carbonyl compounds ≤0.002 % (Acetone)
Titrable acid or base ≤0.0001 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR