Ethyl-2-(triphenylphosphoranylidene)acetate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl triphenylphosphoranylidene-acetate, (Triphenylphosphoranylidene)acetic acid ethyl ester, (Carbethoxymethylene)triphenylphosphorane, Ethoxycarbonylmethylene-triphenylphosphorane, Ethyl 2-(triphenylphosphoranylidene)acetate

10829-0250 10829-1000 10829-5000
TS10829-0250EA 95 USD
TS10829-0250 TS10829-1000 TS10829-5000
Ethyl-2-(triphenylphosphoranylidene)acetate 98+%
Ethyl-2-(triphenylphosphoranylidene)acetate
Formula: C₂₂H₂₁O₂P
MW: 348.38 g/mol
Melting Pt: 124…129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009183
CAS Number: 1099-45-2

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 124 to 129 °C
HPLC ≥98.0 %
UV at 250 nm A: ≤0.05
Titration after Ion exchange ≥97.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR