Butyraldehyde 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Butylaldehyde, Butanal

10809-0010 10809-0025 10809-1000
TS10809-0010EA 62 USD
TS10809-0010 TS10809-0025 TS10809-1000
Butyraldehyde 99%
Butyraldehyde
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 75 °C (760 mmHg)
Melting Pt: –96 °C
Density: 0.817 g/cm³
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00007023
CAS Number: 123-72-8
EINECS: 204-646-6
UN: 1129
ADR: 3 II
Merck Index: 1301591

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3770 to 1.3830 (20°C, 589 nm)
GC ≥98.5 %
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR