Acetophenone 98%, pure

Supplier: Thermo Scientific Chemicals
Warning

10241-0010 10241-0025 10241-0050 10241-0100 10241-2500
TS10241-0010EA 74 USD
TS10241-0010 TS10241-0025 TS10241-0050 TS10241-0100 TS10241-2500
Acetophenone 98%, pure
Acetophenone
Formula: C₈H₈O
Boiling Pt: 202 °C (760 mmHg)
Melting Pt: 19.6 °C
Density: 1.028 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00008724
CAS Number: 98-86-2
EINECS: 202-708-7
Merck Index: 1300074

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5350 (20°C, 589 nm)
GC ≥97.5 %
Visual transition interval from pH 9.4 (orange-brown) to pH 12.0 (violet)
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR