3-Hydroxytyrosol ≥98% (by HPLC), Sigma-Aldrich®

Warning

Synonyms: 2-(3,4-Dihydroxyphenyl)ethanol, Hydroxytyrosol, DOPET, Homoprotocatechuyl alcohol, 2-(3,4-Dihydroxyphenyl)ethyl alcohol, 3,4-Dihydroxyphenethyl Alcohol

M-Clarity™ quality level = MQ100

H4291-25MG H4291-100MG
220182-148EA 369.23 USD
220182-148 220182-147
3-Hydroxytyrosol ≥98% (by HPLC), Sigma-Aldrich®
3-Hydroxytyrosol

Metabolite of oleuropein. Antioxidant. Inhibits the rate of cancer cell proliferation and induces cancer cell apoptosis.

Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01320529
CAS Number: 10597-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR