3,3'-Iminobis(N,N-dimethylpropylamine)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N'-[3-(Dimethylamino)propyl]-N,N-dimethyl-1,3-propanediamine, N1-(3-(Dimethylamino)propyl)-N3,N3-dimethylpropane-1,3-diamine, N,N-Bis[3-(dimethylamino)propyl]amine

46634-0250 46634-1000 46634-5000
TS46634-0250EA 27 USD
TS46634-0250 TS46634-1000 TS46634-5000
3,3'-Iminobis(N,N-dimethylpropylamine)
3,3'-Iminobis(N,N-dimethylpropylamine)
Formula: C₁₀H₂₅N₃
MW: 187.33 g/mol
Boiling Pt: 128.0°C ...131.0°C (27.0 hPa)
Density: 0.8410 g/mL
Flash Pt: 98°C
MDL Number: MFCD00014880
CAS Number: 6711-48-4
EINECS: 229-761-9
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to brown liquid
Infrared spectrum Conforms
Refractive index 1.4470 to 1.4510 (20°C, 589 nm)
Titration Total base 96.0 to 104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR