Casamino Acids, Powder, Thermo Scientific
Supplier: Thermo Scientific Chemicals
61204-1000
61204-5000
TS61204-1000EA
137.91
USD
TS61204-1000
TS61204-5000
Casamino Acids, Powder, Thermo Scientific
Media for Microbiology
Dehydrated Media for Microbiology
MDL No.: MFCD00217199
Storage Temperature: Ambient
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...