Diethyl ether, anhydrous ACS

Supplier: Thermo Scientific Chemicals
Danger

61508-0010 61508-0040 61508-5000
TS61508-0010EA 121 USD
TS61508-0010 TS61508-0040 TS61508-5000
Diethyl ether, anhydrous ACS
Diethyl ether
Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: –116 °C
Density: 0.714 g/cm³ (20 °C)
Flash Pt: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
UN: 1155
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=9e4ec8d25df2fecee88827e09071c0f955486274&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Assay ≥99.0 %
Water ≤0.01 %
Density (25 °C) ≤0.7079
Additional info BHT: report (About 7.5 ppm)
Color scale ≤10 APHA
Identification passes test
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 passes test
Copper (Cu) ≤0.1 ppm
Nickel (Ni) ≤0.1 ppm
Heavy metals (as Pb) ≤1 ppm
Carbonyl (as HCHO) ≤0.001 %
Titrable acid ≤0.0002 meq/g
Peroxides ≤1 ppm (as H2O2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR