Aphidicolin

Supplier: Thermo Scientific Chemicals

Synonyms: (3R,4R,4aR,6aS,8R,9R,11aS,11bS)-4,9-Bis(hydroxymethyl)-4,11b-dimethyltetradecahydro-8,11a-methanocyclohepta[a]naphthalene-3,9-diol

61197-0050 61197-0010
TS61197-0050EA 424 USD
TS61197-0050 TS61197-0010
Aphidicolin
Aphidicolin
Formula: C₂₀H₃₄O₄
MW: 338.488 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00083214
CAS Number: 38966-21-1

Order Now

Specification Test Results

Appearance White to off-white Solid
Melting point 226 to 235 °C
Solubility (1 % in methanol) Passes test
TLC ≥98 %
Identification Passes test
Elemental Analysis 9.92 to 10.32 % (H)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR