Phenol : chloroform : iso-amyl alcohol (25:24:1) saturated with 100 mM Tris-EDTA to pH 8.0 stabilized for molecular biology DNase-, RNase-, protease-free pH 7.8-8.2
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Carbolic acid, Hydroxybenzene, Phenic acid
32711-5000
32711-0025
32711-1000
TS32711-1000EA
104.72
USD
TS32711-1000
TS32711-0025
TS32711-5000
Phenol : chloroform : iso-amyl alcohol (25:24:1) saturated with 100 mM Tris-EDTA to pH 8.0 stabilized for molecular biology DNase-, RNase-, protease-free pH 7.8-8.2
Phenol : Chloroform : iso-Amyl alcohol
Density:
1.28 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00133763 CAS Number: 136112-00-0 UN: 2810 ADR: 6.1,II |
Specification Test Results
Appearance | Clear yellow to orange Solution |
Stabilizer | 0.08 to 0.12 % hydroxyquinoline |
pH | 7.8 to 8.2 (at 25°C) |
Composition | mixture of Phenol/Chloroform/Isoamyl alcohol in the ratio of 25:24:1 (v/v/v) |
Dnase,Rnase,Protease activity | None detected |
Iron (Fe) | ≤1 ppm |
Heavy metals (as Pb) | ≤1 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...