Mineral oil (petroleum), pure
Supplier: Thermo Scientific Chemicals
41508-0100
41508-0010
41508-0025
41508-0060
TS41508-0100EA
360
USD
TS41508-0100
TS41508-0010
TS41508-0025
TS41508-0060
Mineral oil (petroleum), pure
Mineral oil (petroleum)
Density:
0.877 g/cm³ Flash Pt: 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00131611 CAS Number: 8042-47-5 |
Specification Test Results
Appearance | Clear colorless Oily liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4730 to 1.4800 (20°C, 589 nm) |
Density | (15°C) 0.860 to 0.880 g/ml |
Viscosity | 63 to 80 mm²/s (40°C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...