Petroleum spirit, 100…140 °C, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
23302-0025
233020100
233020250
TS23302-0025EA
164
USD
TS23302-0025
TS233020100
TS233020250
Petroleum spirit, 100…140 °C, extra pure
Petroleum spirit
Boiling Pt:
100…140 °C (760 mmHg) Melting Pt: –30 °C Density: 0.74 g/cm³ Flash Pt: 2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081849 CAS Number: 64742-49-0 EINECS: 265-151-9 UN: 3295 ADR: 3,II |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4070 to 1.4150 (20°C, 589 nm) |
Boiling point | 98 to 140°C |
Density | (15°C) 0.737 to 0.747 g/ml |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.005 % |
Benzene | ≤50 ppm |
Aromatics | ≤0.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...