Silica gel C18-RP 17% C (∼0.7 mmol/g) Ø 48-65 µm

Supplier: Thermo Scientific Chemicals

36012-0500 360122500
TS36012-0500EA 479 USD
TS36012-0500 TS360122500
Silica gel C18-RP 17% C (∼0.7 mmol/g) Ø 48-65 µm
Silica gel C18-RP
Storage Temperature: Ambient
MDL Number: MFCD00241275
CAS Number: 71889-02-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Particle size 48.2 to 65.3 µm (D50)
Loading ≥0.67 mmol/g (Molecular)
Specific surface area 500 to 600 m²/g
Pore volume 0.80 to 1.00 ml/g
Carbon (C) ≥16 %
Volatile matter ≤4 % (110°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR