Silica gel C18-RP 23% C (∼1 mmol/g) Ø 40-63 µm

Supplier: Thermo Scientific Chemicals

36011-2500 360110500
TS36011-2500EA 1428 USD
TS36011-2500 TS360110500
Silica gel C18-RP 23% C (∼1 mmol/g) Ø 40-63 µm
Silica gel C18-RP
Storage Temperature: Ambient
MDL Number: MFCD00241275
CAS Number: 71889-02-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Particle size 40 to 63 µm
Loading ≥0.96 mmol/g (Molecular)
Specific surface area 480 to 550 m²/g
Pore volume 0.70 to 0.85 ml/g
Carbon (C) ≥20.7 %
Volatile matter ≤4 % (110°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR