Quartz, coarse wool 9-30 µm

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Silicium dioxide, Silicium (IV) oxide, Silicon dioxide, Sand, white quartz

39361-1000 393610100
TS39361-1000EA 317 USD
TS39361-1000 TS393610100
Quartz, coarse wool 9-30 µm
Quartz
Formula: SiO₂
MW: 60.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00217788
CAS Number: 14808-60-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5273273c3b2e473c6ced4e97e18a338f3d21302c&vid=9c414ebb18d830bfa8a77b318dbbb0e6a96da07b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Coarse fibers

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR