Silica gel, Pore size: 60 Å 0.200-0.500 mm for chromatography
Supplier: Thermo Scientific Chemicals
24038-0050
240380010
TS24038-0050EA
484
USD
TS24038-0050
TS240380010
Silica gel, Pore size: 60 Å 0.200-0.500 mm for chromatography
Silica gel
Formula:
SiO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 REACH: 01-2119379499-16 |
Specification Test Results
Appearance | White Powder |
Particle size | ≤10 % (undersized particles) |
Specific surface area | 480 to 550 m²/g |
Pore volume | 0.55 to 0.85 ml/g |
pH | 6.0 to 7.5 (5 % suspension in water) |
Residual water | ≤4 % (150°C, 2 h) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...